Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Tigloylglycine 98.0+%, TCI America™
SDP

Catalog No. T1260100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1260100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1260100MG Supplier TCI America Supplier No. T1260100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Unnatural amino acid derivative for use in peptide chemistry research.

Chemical Identifiers

CAS 35842-45-6
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD00059118
InChI Key WRUSVQOKJIDBLP-HWKANZROSA-N
PubChem CID 6441567
ChEBI CHEBI:73018
IUPAC Name 2-[[(E)-2-methylbut-2-enoyl]amino]acetic acid
SMILES CC=C(C)C(=O)NCC(=O)O

Specifications

Melting Point 86°C
Color White-Yellow
Packaging Glass Bottle with Plastic Insert
Quantity 100 mg
Formula Weight 157.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Tigloylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.