accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(S)NH2OOHCl

CAS RN 35858-81-2

IUPAC Name: benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Synonyms: benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Molecular Weight (g/mol): 329.8
Molecular Formula: C18H18ClN2O2
InChi Key: BYXCCQMIJIPTSL-UHFFFAOYNA-N
SMILES: [Cl].NC(CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)