Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tryptophan benzyl ester hydrochloride, 98%, Thermo Scientific™

Catalog No. AAH6338506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6338506 5 g
AAH6338503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6338506 Supplier Thermo Scientific Chemicals Supplier No. H6338506

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-Tryptophan benzyl ester hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol (50 mg/mL in MeOH).

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35858-81-2
Molecular Formula C18H18ClN2O2
Molecular Weight (g/mol) 329.80
MDL Number MFCD00058122
InChI Key BYXCCQMIJIPTSL-UHFFFAOYNA-N
Synonym h-trp-obzl.hcl, l-tryptophan benzyl ester hydrochloride, benzyl l-tryptophanate hydrochloride, h-trp-obzl hcl, h-trp-obzl hydrochloride, s-benzyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-obzlhcl, benzyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, benzyl l-tryptophanate monohydrochloride, c18h18n2o2.hcl
PubChem CID 45073232
SMILES [Cl].NC(CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1

Specifications

Color White
Solubility Information Soluble in methanol (50mg/mL in MeOH).
IUPAC Name benzyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Quantity 5 g
Formula Weight 330.81
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Tryptophan benzyl ester hydrochloride
EINECSNumber 252-765-7
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.