accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3587-62-0

IUPAC Name: (chloromethoxy)cyclohexane
Synonyms: (chloromethoxy)cyclohexane
Molecular Weight (g/mol): 148.63
Molecular Formula: C7H13ClO
InChi Key: GYCLGFVWNVHUPA-UHFFFAOYSA-N
SMILES: ClCOC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results