CAS RN 35895-69-3
CAS RN 35895-69-3
IUPAC Name:
tetraethylazanium trifluoromethanesulfonate
Synonyms:
tetraethylammonium triflate
Molecular Weight (g/mol):
279.32
Molecular Formula:
C9H20F3NO3S
InChi Key:
PUZYNDBTWXJXKN-UHFFFAOYSA-M
SMILES:
[O-]S(=O)(=O)C(F)(F)F.CC[N+](CC)(CC)CC
1
–
1
of
1
results
1
Tetraethylammonium Trifluoromethanesulfonate 99.0+%, TCI America™
CAS: 35895-69-3 Molecular Formula: C9H20F3NO3S Molecular Weight (g/mol): 279.318 MDL Number: MFCD00042586 InChI Key: PUZYNDBTWXJXKN-UHFFFAOYSA-M Synonym: Tetraethylammonium Triflate PubChem CID: 2776566 IUPAC Name: tetraethylazanium;trifluoromethanesulfonate SMILES: CC[N+](CC)(CC)CC.C(F)(F)(F)S(=O)(=O)[O-]