accessibility menu, dialog, popup

UserName

Viewing profile as user:
SClCl

CAS RN 3592-44-7

IUPAC Name: chloro[(chloromethyl)sulfanyl]methane
Synonyms: chloro[(chloromethyl)sulfanyl]methane
Molecular Weight (g/mol): 131.01
Molecular Formula: C2H4Cl2S
InChi Key: KRIFIIWBVJKVST-UHFFFAOYSA-N
SMILES: ClCSCCl
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results