accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHF

CAS RN 359446-04-1

IUPAC Name: [(4-fluorophenyl)methyl](2-methylpropyl)amine
Synonyms: [(4-fluorophenyl)methyl](2-methylpropyl)amine
Molecular Weight (g/mol): 181.25
Molecular Formula: C11H16FN
InChi Key: SOUYVJATMXDMQK-UHFFFAOYSA-N
SMILES: CC(C)CNCC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results