accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35963-20-3

IUPAC Name: [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Synonyms: [(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Molecular Weight (g/mol): 232.29
Molecular Formula: C10H16O4S
InChi Key: MIOPJNTWMNEORI-MHPPCMCBSA-N
SMILES: CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
Molecular Weight (g/mol) 
  • (10)
  • (3)
Quantity 
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (7)
  • (10)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (7)