Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Vinyltrimethylsilane

Catalog No. 21395025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-395-025G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 21-395-025G Supplier Sigma Aldrich Supplier No. 21395025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used for the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.

Specifications

CAS 35963-20-3
Molecular Formula C10H16O4S
Refractive Index n20/D 1.391
Linear Formula C10H16O4S
MDL Number MFCD00064158
Quantity 250 g
Synonym Trimethyl(vinyl)silane; (Trimethylsilyl)ethylene; Ethenyltrimethylsilane
Molecular Weight (g/mol) 232.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.