accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NSCH3OOHOOHO(E)

CAS RN 3598-37-6

IUPAC Name: (2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
Synonyms: acepromazine
Molecular Weight (g/mol): 442.53
Molecular Formula: C23H26N2O5S
InChi Key: FQRHOOHLUYHMGG-WLHGVMLRSA-N
SMILES: OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (6)
Percent Purity 
  • (6)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (6)
  • (6)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (6)
  • (6)