Learn More
Acepromazine Maleate, USP, 98-101%, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.2% |
| Melting Range | 136°C to 139°C |
| Packaging | Amber Glass Bottle |
| Quantity | 1 kg |
| Percent Purity | 98 to 101% |
| Grade | USP |
| Physical Form | Crystals or Flakes |
| Chemical Name or Material | Acepromazine Maleate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.