accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CClFeClClCl

CAS RN 359845-21-9

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium; tetrachloroironuide
Synonyms: 1-butyl-3-methylimidazolium; tetrachloroferrate(1-)
Molecular Weight (g/mol): 336.87
Molecular Formula: C8H15Cl4FeN2
InChi Key: FYHLFBVHUIJIII-UHFFFAOYSA-J
SMILES: Cl[Fe-](Cl)(Cl)Cl.CCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results