Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Tetrachloroferrate 98.0+%, TCI America™
SDP

Catalog No. B26725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26725G 5 g
B267225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26725G Supplier TCI America Supplier No. B26725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 359845-21-9
Molecular Formula C8H15Cl4FeN2
Molecular Weight (g/mol) 336.867
InChI Key FYHLFBVHUIJIII-UHFFFAOYSA-J
PubChem CID 11674455
IUPAC Name 1-butyl-3-methylimidazol-3-ium;tetrachloroiron(1-)
SMILES CCCCN1C=C[N+](=C1)C.Cl[Fe-](Cl)(Cl)Cl

Specifications

Color Green-Yellow
Quantity 5 g
Formula Weight 336.87
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Tetrachloroferrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.