accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3599-89-1

IUPAC Name: propanimidamide hydrochloridyl
Synonyms: propamidine hydrochloridyl
Molecular Weight (g/mol): 107.56
Molecular Formula: C3H8ClN2
InChi Key: KBOZRAZQYBTUJV-UHFFFAOYSA-N
SMILES: [Cl].CCC(N)=N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results