Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionamidine Hydrochloride, TCI America™
SDP

Catalog No. P206925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P206925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P206925G Supplier TCI America Supplier No. P206925G

Chemical Identifiers

CAS 3599-89-1
Molecular Formula C3H9ClN2
Molecular Weight (g/mol) 108.569
MDL Number MFCD00233938
InChI Key DFWRZHZPJJAJMX-UHFFFAOYSA-N
Synonym Propanamidine Hydrochloride
PubChem CID 12301253
IUPAC Name propanimidamide;hydrochloride
SMILES CCC(=N)N.Cl

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 108.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Propionamidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.