accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 360-03-2

IUPAC Name: 2-(2,3-difluorophenyl)acetate
Synonyms: 2-(2,3-difluorophenyl)acetate
Molecular Weight (g/mol): 171.12
Molecular Formula: C8H5F2O2
InChi Key: UXSQXUSJGPVOKT-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=CC(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results