accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36042-99-6

IUPAC Name: chlorobis(naphthalen-1-yl)phosphane
Synonyms: chlorobis(naphthalen-1-yl)phosphane
Molecular Weight (g/mol): 320.76
Molecular Formula: C20H14ClP
InChi Key: ZHRMSAAMUUNWKE-UHFFFAOYSA-N
SMILES: ClP(C1=CC=CC2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)