Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodi(1-naphthyl)phosphine, 95%

Catalog No. AAH2589306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2589306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2589306 Supplier Thermo Scientific Chemicals Supplier No. H2589306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36042-99-6 | C20H14ClP | 320.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36042-99-6
Molecular Formula C20H14ClP
Molecular Weight (g/mol) 320.76
MDL Number MFCD09753469
InChI Key ZHRMSAAMUUNWKE-UHFFFAOYSA-N
Synonym chlorodi 1-naphthyl phosphine, bis 1-naphthyl chlorophosphine, phosphinous chloride, di-1-naphthalenyl, chlorobis naphthalen-1-yl phosphane, di 1-naphthyl chlorophosphine, chloro dinaphthalen-1-yl phosphane, dinaphthalen-1-ylphosphinous chloride, phosphinous chloride,p,p-di-1-naphthalenyl
PubChem CID 18705941
SMILES ClP(C1=CC=CC2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 154°C to 157°C
Boiling Point 227°C to 229°C (0.6 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name chlorobis(naphthalen-1-yl)phosphane
Formula Weight 320.76
Percent Purity 95%
Chemical Name or Material Chlorodi(1-naphthyl)phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.