accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOOONNNNO

CAS RN 3605-01-4

IUPAC Name: 2-{4-[(2H-1,3-benzodioxol-5-yl)methyl]piperazin-1-yl}pyrimidine; methanesulfonic acid
Synonyms: piribedil
Molecular Weight (g/mol): 394.45
Molecular Formula: C17H22N4O5S
InChi Key: DEZKFCIOSKCCHK-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.C(N1CCN(CC1)C1=NC=CC=N1)C1=CC=C2OCOC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results