Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piribedil 98.0+%, TCI America™
SDP

Catalog No. P2054200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2054200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2054200MG Supplier TCI America Supplier No. P2054200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 3605-01-4
Melting Point 98°C
Color White
Molecular Formula C17H22N4O5S
Quantity 200 mg
MDL Number MFCD00868264
Synonym 2-[4-(1,3-Benzodioxol-5-ylmethyl)-1-piperazinyl]pyrimidine
InChI Key DEZKFCIOSKCCHK-UHFFFAOYSA-N
SMILES C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC=CC=N4
IUPAC Name 2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]pyrimidine
Molecular Weight (g/mol) 394.45
PubChem CID 4850
Formula Weight 298.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Piribedil
Show More Show Less
RTECSNumber UV9704000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.