accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36075-06-6

IUPAC Name: N,N-diethylpyridin-2-amine
Synonyms: N,N-diethylpyridin-2-amine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: XRPITCBWOUOJTH-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results