Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diethylaminopyridine 97.0+%, TCI America™
SDP

Catalog No. D16815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16815G 5 g
D168125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16815G Supplier TCI America Supplier No. D16815G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36075-06-6
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.225
MDL Number MFCD00059777
InChI Key XRPITCBWOUOJTH-UHFFFAOYSA-N
Synonym N-(2-Pyridyl)diethylamine
PubChem CID 232542
IUPAC Name N,N-diethylpyridin-2-amine
SMILES CCN(CC)C1=CC=CC=N1

Specifications

Boiling Point 118°C
Quantity 5 g
Formula Weight 150.23
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Diethylaminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.