accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3O

CAS RN 3609-53-8

IUPAC Name: methyl 4-acetylbenzoate
Synonyms: methyl 4-acetylbenzoate
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: QNTSFZXGLAHYLC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)