accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHHON(E/Z)

CAS RN 36145-03-6

IUPAC Name: 3'-hydroxy-1'H,3H-[2,2'-bipyridinyliden]-3-one
Synonyms: 3'-hydroxy-1'H-[2,2'-bipyridinyliden]-3-one
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: ZPHMTLAGULLPGM-UHFFFAOYSA-N
SMILES: OC1=CC=CNC1=C1N=CC=CC1=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results