Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridine-3,3'-diol 98.0+%, TCI America™
SDP

Catalog No. B425550MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B425550MG 50 mg
B4255250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B425550MG Supplier TCI America Supplier No. B425550MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36145-03-6 | C10H8N2O2 | 188.186 g/mol

Chemical Identifiers

CAS 36145-03-6
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00075177
InChI Key ZPHMTLAGULLPGM-KTKRTIGZSA-N
PubChem CID 16212186
IUPAC Name (2Z)-2-(3-hydroxy-1H-pyridin-2-ylidene)pyridin-3-one
SMILES C1=CC(=O)C(=C2C(=CC=CN2)O)N=C1

Specifications

Melting Point 199°C
Quantity 50 mg
Synonym 3,3′C-Dihydroxy-2,2′C-bipyridyl, 3,3′C-Dihydroxy-2,2′C-bipyridine
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-3,3′-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.