accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)OOH(R)OH(S)HO(R)HO

CAS RN 3615-37-0

IUPAC Name: (3R,4S,5R,6R)-6-methyloxane-2,3,4,5-tetrol
Synonyms: D-fucose 6-deoxy-D-galactose D-galactose, 6-deoxy- D-fucopyranose
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: SHZGCJCMOBCMKK-SVZMEOIVSA-N
SMILES: C[C@H]1OC(O)[C@H](O)[C@@H](O)[C@H]1O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)