accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36151-45-8

ONO

CAS RN 36151-45-8

IUPAC Name: 4-(2-oxopyrrolidin-1-yl)benzaldehyde
Synonyms: 4-(2-oxopyrrolidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: VADZUJOCSAESJS-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N1CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results