accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 362-07-2

IUPAC Name: (1S,3aS,3bR,9bS,11aS)-8-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Synonyms: 2-methoxyestradiol 2-(methyl-11c)methoxyestradiol panzem 2-(methyl-11C)methoxyestradiol 2-methoxyestradiol-17 β 2-methoxyestradiol-17β
Molecular Weight (g/mol): 302.41
Molecular Formula: C19H26O3
InChi Key: CQOQDQWUFQDJMK-SSTWWWIQSA-N
SMILES: COC1=CC2=C(CC[C@H]3[C@@H]4CC[C@H](O)[C@@]4(C)CC[C@H]23)C=C1O
Form 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results