Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2-Methoxyestradiol

Catalog No. 45418050MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-418-050MG 50 mg
45-418-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-418-050MG Supplier MilliporeSigma™ Supplier No. 45418050MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A natural metabolite of 17β-estradiol that is devoid of estrogenic activity.

A natural metabolite of 17β-estradiol that is devoid of estrogenic activity. Inhibits cell proliferation and angiogenesis. Markedly inhibits the neovascularization of solid tumors and suppresses tumor growth in mice. Binds to the colchicine binding site of tubulin, and has been suggested to function as a natural regulator of microtubule assembly and function.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Inhibits cell proliferation and angiogenesis
CAS 362-07-2
Molecular Formula C19H26O3
Formula Weight 302.4g/mol
Solubility DMSO or EtOH
Form Crystalline Solid
Quantity 50 mg
Purity 90% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.