accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3627-04-1

IUPAC Name: 4-{2-[4-(diethylamino)-6-oxocyclohexa-2,4-dien-1-ylidene]hydrazin-1-yl}-5-hydroxynaphthalene-2,7-disulfonic acid
Synonyms: 4-{2-[4-(diethylamino)-6-oxocyclohexa-2,4-dien-1-ylidene]hydrazin-1-yl}-5-hydroxynaphthalene-2,7-disulfonic acid
Molecular Weight (g/mol): 495.52
Molecular Formula: C20H21N3O8S2
InChi Key: MXOHFNOCOCYZDR-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC(=O)C(C=C1)=NNC1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results