Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllon III, TCI America™
SDP

Catalog No. B04531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B04531G Supplier TCI America Supplier No. B04531G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3627-04-1
Molecular Formula C20H21N3O8S2
Molecular Weight (g/mol) 495.52
MDL Number MFCD00070629
InChI Key MXOHFNOCOCYZDR-UHFFFAOYSA-N
Synonym 5-(4-Diethylamino-2-hydroxyphenylazo)-4-hydroxynaphthalene-2,7-disulfonic Acid Sodium Salt
PubChem CID 91873463
IUPAC Name 4-[(2Z)-2-[4-(diethylamino)-6-oxocyclohexa-2,4-dien-1-ylidene]hydrazinyl]-5-hydroxynaphthalene-2,7-disulfonic acid
SMILES CCN(CC)C1=CC(=O)C(=NNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C=C1

Specifications

Color Black-Green
Quantity 1 g
Formula Weight 517.50
Physical Form Crystalline Powder
Chemical Name or Material Beryllon III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.