CAS RN 36290-94-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Poly[4-(diacetoxyiodo)styrene], TCI America™
CAS: 36290-94-5 MDL Number: MFCD06798811 Synonym: Polystyrene Iodosoacetate, PSDIB