accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNNOSOOO

CAS RN 36305-05-2

IUPAC Name: 2-methoxy-4-(phenylamino)benzene-1-diazonium sulfate
Synonyms: 2-methoxy-4-(phenylamino)benzenediazonium sulfate
Molecular Weight (g/mol): 322.32
Molecular Formula: C13H12N3O5S
InChi Key: HBDLDBQEQKQUPI-UHFFFAOYSA-L
SMILES: [O-]S([O-])(=O)=O.COC1=CC(NC2=CC=CC=C2)=CC=C1[N+]#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results