Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Diazo-3-methoxydiphenylamine Sulfate 96.0+%, TCI America™
SDP

Catalog No. D228525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D228525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D228525G Supplier TCI America Supplier No. D228525G

Chemical Identifiers

CAS 36305-05-2
Molecular Formula C13H13N3O5S
Molecular Weight (g/mol) 323.323
MDL Number MFCD00083004
InChI Key HBDLDBQEQKQUPI-UHFFFAOYSA-M
Synonym 2-Methoxy-4-(phenylamino)benzenediazonium Hydrogensulfate
PubChem CID 13643126
IUPAC Name 4-anilino-2-methoxybenzenediazonium;hydrogen sulfate
SMILES COC1=C(C=CC(=C1)NC2=CC=CC=C2)[N+]#N.OS(=O)(=O)[O-]

Specifications

Quantity 25 g
Formula Weight 323.32
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diazo-3-methoxydiphenylamine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.