accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3632-91-5

IUPAC Name: magnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Synonyms: magnesium gluconate
Molecular Weight (g/mol): 414.6
Molecular Formula: C12H22MgO14
InChi Key: CTUVIUYTHWPELF-IYEMJOQQSA-L
SMILES: [Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)