accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3634-59-1

IUPAC Name: chloro(hexyl)dimethylsilane
Synonyms: chloro(hexyl)dimethylsilane
Molecular Weight (g/mol): 178.78
Molecular Formula: C8H19ClSi
InChi Key: JQYKSDDVPXVEOL-UHFFFAOYSA-N
SMILES: CCCCCC[Si](C)(C)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results