Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(hexyl)dimethylsilane 96.0+%, TCI America™
SDP

Catalog No. C34155ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34155ML 5 mL
C341525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34155ML Supplier TCI America Supplier No. C34155ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3634-59-1
Molecular Formula C8H19ClSi
Molecular Weight (g/mol) 178.78
MDL Number MFCD01073817
InChI Key JQYKSDDVPXVEOL-UHFFFAOYSA-N
Synonym chloro hexyl dimethylsilane, hexyldimethylchlorosilane, dimethylhexylsilyl chloride, hexylchlorodimethylsilane, hexyldi-methylchlorosilane, dimethyl hexylsilyl chloride
PubChem CID 2733540
IUPAC Name chloro(hexyl)dimethylsilane
SMILES CCCCCC[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 184°C
Quantity 5 mL
UN Number 2987
Formula Weight 178.78
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(hexyl)dimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.