CAS RN 36362-97-7
IUPAC Name:
benzene-1,2,3-tricarboxylate
Synonyms:
benzene-1,2,3-tricarboxylate
Molecular Weight (g/mol):
207.12
Molecular Formula:
C9H3O6
InChi Key:
UJMDYLWCYJJYMO-UHFFFAOYSA-K
SMILES:
[O-]C(=O)C1=CC=CC(C([O-])=O)=C1C([O-])=O
1
–
1
of
1
results
1
1,2,3-Benzenetricarboxylic Acid Hydrate 96%, Thermo Scientific™
CAS: 36362-97-7 Molecular Formula: C9H3O6 Molecular Weight (g/mol): 207.12 MDL Number: MFCD00149097 InChI Key: UJMDYLWCYJJYMO-UHFFFAOYSA-K Synonym: 1,2,3-benzenetricarboxylic acid hydrate dihydrate PubChem CID: 118797571 IUPAC Name: benzene-1,2,3-tricarboxylate SMILES: [O-]C(=O)C1=CC=CC(C([O-])=O)=C1C([O-])=O