Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Benzenetricarboxylic Acid Hydrate 96%, Thermo Scientific™

Catalog No. AC105340250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105340250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105340250 Supplier Thermo Scientific Chemicals Supplier No. 105340250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36362-97-7
Molecular Formula C9H3O6
Molecular Weight (g/mol) 207.12
MDL Number MFCD00149097
InChI Key UJMDYLWCYJJYMO-UHFFFAOYSA-K
Synonym 1,2,3-benzenetricarboxylic acid hydrate dihydrate
PubChem CID 118797571
IUPAC Name benzene-1,2,3-tricarboxylate
SMILES [O-]C(=O)C1=CC=CC(C([O-])=O)=C1C([O-])=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 192°C to 198°C
Color White
Infrared Spectrum Authentic
Linear Formula C6H3(CO2H)3.xH2O
Quantity 25 g
Beilstein 09, 976
Formula Weight 210.14
Percent Purity ≥95% (on dry substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2, 3-Benzenetricarboxylic acid hydrate, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.