accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3639-21-2

CH3OHCH3OO

CAS RN 3639-21-2

IUPAC Name: 2-ethyl-2-hydroxybutanoate
Synonyms: 2-ethyl-2-hydroxybutanoate
Molecular Weight (g/mol): 131.15
Molecular Formula: C6H11O3
InChi Key: LXVSANCQXSSLPA-UHFFFAOYSA-M
SMILES: CCC(O)(CC)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results