accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36436-65-4

IUPAC Name: 1-(2-hydroxy-4,5-dimethylphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-4,5-dimethylphenyl)ethanone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: YXVSURZEXVMUAM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=C(C)C=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)