accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3644-12-0

IUPAC Name: N-(methoxymethyl)-2-methylprop-2-enamide
Synonyms: N-(methoxymethyl)-2-methylprop-2-enamide
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: YOZHLACIXDCHPV-UHFFFAOYSA-N
SMILES: COCNC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results