Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Methoxymethyl)methacrylamide (stabilized with MEHQ) 85.0+%, TCI America™
SDP

Catalog No. M2478500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2478500G 500 g
M247825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2478500G Supplier TCI America Supplier No. M2478500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3644-12-0
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00035808
InChI Key YOZHLACIXDCHPV-UHFFFAOYSA-N
PubChem CID 77204
IUPAC Name N-(methoxymethyl)-2-methylprop-2-enamide
SMILES COCNC(=O)C(C)=C

Specifications

Color Yellow
Boiling Point 85°C
Quantity 500 g
Formula Weight 129.16
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(Methoxymethyl)methacrylamide (stabilized with MEHQ)
EINECSNumber (2)-1067
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.