accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3644-56-2

IUPAC Name: 2-chloro-N-(2,6-dichlorophenyl)acetamide
Synonyms: 2-chloro-N-(2,6-dichlorophenyl)acetamide
Molecular Weight (g/mol): 238.49
Molecular Formula: C8H6Cl3NO
InChi Key: CZAFLRAOEDDMCS-UHFFFAOYSA-N
SMILES: ClCC(=O)NC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results