Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-N-(2,6-dichlorophenyl)-acetamide, 99%, Thermo Scientific™

Catalog No. AC125041000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC125041000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC125041000 Supplier Thermo Scientific Chemicals Supplier No. 125041000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3644-56-2
Molecular Formula C8H6Cl3NO
Molecular Weight (g/mol) 238.49
MDL Number MFCD00044773
InChI Key CZAFLRAOEDDMCS-UHFFFAOYSA-N
Synonym 2-chloro-n-2,6-dichlorophenyl acetamide, n-2,6-dichlorophenyl-2-chloroacetamide, acetamide,2-chloro-n-2,6-dichlorophenyl, 2,6-dichlorophenyl aminocarbonylmethyl chloride, n-2,6-dichlorophenyl chloroacetamide, 2-chloro-n-2,6-dichlorophenyl-acetamide, 2-chloro-n-2,6-dichloro-phenyl-acetamide
PubChem CID 77206
IUPAC Name 2-chloro-N-(2,6-dichlorophenyl)acetamide
SMILES ClCC(=O)NC1=C(Cl)C=CC=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Quantity 100 g
Formula Weight 238.5
Chemical Name or Material 2-chloro-N-(2, 6-dichlorophenyl)-acetamide, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.