accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3647-74-3

IUPAC Name: 4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: 4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: GPIUUMROPXDNRH-UHFFFAOYNA-N
SMILES: O=C1NC(=O)C2C3CC(C=C3)C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results