Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Norbornene-2,3-dicarboximide 98.0+%, TCI America™
SDP

Catalog No. N090325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N090325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N090325G Supplier TCI America Supplier No. N090325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3647-74-3
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD08704199
InChI Key GPIUUMROPXDNRH-UHFFFAOYNA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2,3-dicarboximide
PubChem CID 97723
IUPAC Name 4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES O=C1NC(=O)C2C3CC(C=C3)C12

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2,3-dicarboximide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.