accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36499-49-7

IUPAC Name: 3-[4-(diethylamino)-2-methylphenyl]-3-(1,2-dimethyl-1H-indol-3-yl)-1,3-dihydro-2-benzofuran-1-one
Synonyms: 3-[4-(diethylamino)-2-methylphenyl]-3-(1,2-dimethylindol-3-yl)-2-benzofuran-1-one
Molecular Weight (g/mol): 438.57
Molecular Formula: C29H30N2O2
InChi Key: LCGJEUAHECYSFW-UHFFFAOYNA-N
SMILES: CCN(CC)C1=CC(C)=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=C(C)N(C)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results