accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 365-12-8

IUPAC Name: 2'-fluoro-[1,1'-biphenyl]-4-carboxylate
Synonyms: 2'-fluoro-[1,1'-biphenyl]-4-carboxylate
Molecular Weight (g/mol): 215.2
Molecular Formula: C13H8FO2
InChi Key: SLKZDWAZOKIEEU-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results