accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NO

CAS RN 3651-67-0

IUPAC Name: hydrogen 4-methylmorpholine chloride
Synonyms: hydrogen N-methylmorpholine chloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: BTJRKNUKPQBLAL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)